
IGEPAL® CA-630

Synonym(s):
Octylphenoxy poly(ethyleneoxy)ethanol, branched
Linear Formula:
(C2H4O)nC14H22O
CAS Number:
Products
Product Category
Brand
Feature
Physical Form
Quality Segment
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing