Skip to Content
Merck
Homoplantaginin ≥98% (HPLC)

Homoplantaginin

Homoplantaginin ≥98% (HPLC)
Synonym(s):
4′,5,7-Trihydroxy 6-methoxyflavone 7-glucoside, 6-Methoxyapigenin 7-O-β-D-glucoside, 6-Methoxyapigenin 7-O-glucoside, 6-Methylscutellarein 7-glucoside, 7-(β-D-Glucopyranosyloxy)-5-hydroxy-2-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, Dinatin 7-glucoside, Hispidulin 7-β-glucoside, Hispiduloside, Salvitin 7-glucoside, Scutellarein 6-methyl ether 7-glucoside
Empirical Formula (Hill Notation):
C22H22O11
CAS Number:
Molecular Weight:
462.40

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing