Gymnemagenin
Synonym(s):
Olean-12-ene-3β,16β,21β,22α,23,28-hexol
Empirical Formula (Hill Notation):
C30H50O6
CAS Number:
Molecular Weight:
506.71
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3