Skip to Content
Merck
Guanosine 5′-triphosphate sodium salt hydrate ≥90% (HPLC)

Guanosine 5′-triphosphate sodium salt hydrate

Guanosine 5′-triphosphate sodium salt hydrate ≥90% (HPLC)
Synonym(s):
5′-GTP-Na2
Empirical Formula (Hill Notation):
C10H16N5O14P3 · xNa+ · yH2O
CAS Number:
Molecular Weight:
523.18 (anhydrous free acid basis)
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing