
Ginsenoside C-K

Synonym(s):
Ginsenoside Compound K, (3B,12B)-3,12-Dihydroxydammar-24-en-20-yl β-D-glucopyranoside, 20(S)-Ginsenoside Ck, Ginsenoside K, Protopanaxadiol 20-O-glucoside
Empirical Formula (Hill Notation):
C36H62O8
CAS Number:
Molecular Weight:
622.87
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing