
Epoprostenol sodium

Synonym(s):
Prostaglandin I2 sodium salt, (5Z,9α,11α,13E,15S)-6,9-Epoxy-11,15-dihydroxyprosta-5,13-dien-1-oic acid sodium salt, Epoprostenol sodium salt, PGI2-Na, Prostacyclin sodium salt
Empirical Formula (Hill Notation):
C20H31NaO5
CAS Number:
Molecular Weight:
374.45
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing