
Doxazosin Related Compound A

Synonym(s):
N-(1,4-Benzodioxane-2-carbonyl)piperazine
Empirical Formula (Hill Notation):
C13H16N2O3 · HCl
CAS Number:
Molecular Weight:
284.74
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2