Skip to Content
Merck
Dorzolamide for system suitability European Pharmacopoeia (EP) Reference Standard

Dorzolamide for system suitability

Dorzolamide for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Dorzolamide hydrochloride
Empirical Formula (Hill Notation):
C10H16N2O4S3 · HCl
CAS Number:
Molecular Weight:
360.90

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1