Skip to Content
BT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DL-Serine
Synonym(s):
(±)-2-Amino-3-hydroxypropionic acid, DL-Serine
Linear Formula:
HOCH
2
CH(NH
2
)COOH
CAS Number:
302-84-1
Molecular Weight:
105.09
EC Number:
206-130-6
Products
Product Category
amino acids (2)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Amino Acid
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S4375
DL-Serine,
≥98% (TLC)
Expand
View Pricing
8.41235
DL-Serine,
for synthesis
Expand
View Pricing
S5386
DL-Serine,
BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Expand
View Pricing