![(±)-3,4-Methylenedioxymethamphetamine hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/323/752/f20ff95e-82ee-40c9-ad8a-1411cf7a650d/640/f20ff95e-82ee-40c9-ad8a-1411cf7a650d.png)
(±)-3,4-Methylenedioxymethamphetamine hydrochloride
![(±)-3,4-Methylenedioxymethamphetamine hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/323/752/f20ff95e-82ee-40c9-ad8a-1411cf7a650d/640/f20ff95e-82ee-40c9-ad8a-1411cf7a650d.png)
Synonym(s):
(±)-MDMA hydrochloride, DL-3,4-Methylenedioxymethamphetamine, DL-MDMA hydrochloride, Ecstasy, XTC hydrochloride
Empirical Formula (Hill Notation):
C11H15NO2 · HCl
CAS Number:
Molecular Weight:
229.70
Products
Product Category
Brand
Formula Weight
Research Area
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing