
Diltiazem for system suitability

Synonym(s):
(+)-cis-Diltiazem hydrochloride, (2S,3S)-(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, CRD-401
Empirical Formula (Hill Notation):
C22H26N2O4S · HCl
CAS Number:
Molecular Weight:
450.98
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing