
Diisobutyl phthalate-3,4,5,6-d4

Empirical Formula (Hill Notation):
C16D4H18O4
CAS Number:
Molecular Weight:
282.37
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing