
Dihydroergotamine mesilate

Synonym(s):
Dihydroergotamine methanesulfonate salt
Empirical Formula (Hill Notation):
C33H37N5O5 · CH4O3S
CAS Number:
Molecular Weight:
679.78
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing