
Dibenzosuberone

Synonym(s):
Dibenzosuberone, 5-Dibenzosuberone, 10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-one, Dibenzocycloheptadienone
Empirical Formula (Hill Notation):
C15H12O
CAS Number:
Molecular Weight:
208.26
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3