
Diallyl disulfide

Synonym(s):
Allyl disulfide, Diallyl disulfide
Linear Formula:
CH2=CHCH2SSCH2CH=CH2
CAS Number:
Molecular Weight:
146.27
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Color
Formula Weight
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing