
Cyclopropyltriphenylphosphonium bromide

Synonym(s):
NSC 110599
Linear Formula:
C3H5P(C6H5)3Br
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing