
Cyclopentadienyllithium

Synonym(s):
1,3-Cyclopentadiene lithium complex, Lithium cyclopentadienide, Lithium cyclopentadienylide
Empirical Formula (Hill Notation):
C5H5Li
CAS Number:
Molecular Weight:
72.03
Products
Product Category
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing