Skip to Content
Merck
Curcumin matrix substance for MALDI-MS, ≥99.5% (HPLC)

Curcumin

Curcumin matrix substance for MALDI-MS, ≥99.5% (HPLC)
Synonym(s):
(E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Diferuloylmethane, Diferulylmethane, Natural Yellow 3, Curcumin
Linear Formula:
[HOC6H3(OCH3)CH=CHCO]2CH2
CAS Number:
Molecular Weight:
368.38
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique