
Concanamycin A

Synonym(s):
Folimycin
Empirical Formula (Hill Notation):
C46H75NO14
CAS Number:
Molecular Weight:
866.09
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing