Chlorprothixene hydrochloride
Synonym(s):
2-Chloro-9-(3-dimethylaminopropylidene)thioxanthene hydrochloride
Empirical Formula (Hill Notation):
C18H18ClNS · HCl
CAS Number:
Molecular Weight:
352.32
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing