
Carvedilol Related Compound D

Synonym(s):
4-(2-Oxiranylmethoxy)-9H-carbazole, 4-(2,3-Epoxypropoxy)carbazole, 4-(Oxiran-2-ylmethoxy)-9H-carbazole
Empirical Formula (Hill Notation):
C15H13NO2
CAS Number:
Molecular Weight:
239.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2