
Carmustine Related Compound A

Synonym(s):
1,3-Bis(2-chloroethyl)urea, BCU, NSC 36198
Empirical Formula (Hill Notation):
C5H10Cl2N2O
CAS Number:
Molecular Weight:
185.05
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2