Skip to Content
BT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calmagite
Synonym(s):
1-(1-Hydroxy-4-methyl-2-phenylazo)-2-naphthol-4-sulfonic acid, 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)naphthalene-1-sulfonic acid
Linear Formula:
HOC
10
H
5
[N=NC
6
H
3
(OH)CH
3
]SO
3
H
CAS Number:
3147-14-6
Molecular Weight:
358.37
EC Number:
221-563-0
Products
Brand
Supelco (1)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Quality Segment
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C204
Calmagite,
metal indicator
Expand
View Pricing