
Bromuconazole

Synonym(s):
1-[[4-Bromo-2-(2,4-dichlorophenyl)tetrahydro-2-furanyl]methyl]- 1H-1,2,4-triazole
Empirical Formula (Hill Notation):
C13H12BrCl2N3O
CAS Number:
Molecular Weight:
377.06
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique