
Barium titanate(IV)

Synonym(s):
Titanium barium oxide, Barium titanate (IV)
Linear Formula:
BaTiO3
CAS Number:
Molecular Weight:
233.19
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Core
Showing 1-5 of 5
Compare
Product Number
Description
Pricing