
Azacitidine Related Compound A

Synonym(s):
5-Azacytosine, 2-Hydroxy-4-aminotriazine, 4-Amino-1,3,5-triazin-2-ol, 4-Amino-1,3,5-triazin-2-one, 4-Amino-2-hydroxy-1,3,5-triazine, NSC 51100, NSC 54006
Empirical Formula (Hill Notation):
C3H4N4O
CAS Number:
Molecular Weight:
112.09
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2