Atazanavir sulfate
Synonym(s):
Dimethyl (3S,8S,9S,12S)-9-benzyl-3,12,di-tert-butyl-8-hydroxy-4,11-dioxo-6-(p-2-pyridylbenzyl)-2,5,6,10,13-pentaazatetradecanedioate sulfate (1:1), Atazanavir sulfate, (3S,8S,9S,12S)-1,14-Dimethyl-3,12-bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-pentaazatetradecanedioate sulfate
Empirical Formula (Hill Notation):
C38H52N6O7 · H2SO4
CAS Number:
Molecular Weight:
802.93
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing