
Apyrase from potatoes

Synonym(s):
Adenosine 5′-diphosphatase, Adenosine 5′-triphosphatase, Apyrase from potatoes
CAS Number:
Products
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-6 of 6
Compare
Product Number
Description
Pricing