Anti-Mouse IgG (Fab specific) F(ab′)2 fragment–FITC antibody produced in goat
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing