Skip to Content
Merck
Anti-Interleukin-8 antibody produced in goat affinity isolated antibody, lyophilized powder

Anti-Interleukin-8 antibody produced in goat

Anti-Interleukin-8 antibody produced in goat affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-CXCL8, Anti-IL-8

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-1 of 1