
Anti-Goat Serum antibody produced in rabbit

Synonym(s):
Anti-Serum, goat
Products
Product Category
Brand
Biological Source
Clonality
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing