
Ammonium molybdate tetrahydrate

Synonym(s):
Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate
Linear Formula:
(NH4)6Mo7O24 · 4H2O
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing