![5-(N-Ethyl-N-isopropyl)amiloride](/deepweb/assets/sigmaaldrich/product/structures/217/711/4a60d4ff-722b-464f-b276-47030ac043a0/640/4a60d4ff-722b-464f-b276-47030ac043a0.png)
5-(N-Ethyl-N-isopropyl)amiloride
![5-(N-Ethyl-N-isopropyl)amiloride](/deepweb/assets/sigmaaldrich/product/structures/217/711/4a60d4ff-722b-464f-b276-47030ac043a0/640/4a60d4ff-722b-464f-b276-47030ac043a0.png)
Synonym(s):
EIPA
Empirical Formula (Hill Notation):
C11H18ClN7O
CAS Number:
Molecular Weight:
299.76
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing