Skip to Content
Merck
4-Aminophenylboronic acid hydrochloride 95%

4-Aminophenylboronic acid hydrochloride

4-Aminophenylboronic acid hydrochloride 95%
Linear Formula:
C6H8BNO2 · HCl
CAS Number:
Molecular Weight:
173.41

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing