
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt

Synonym(s):
5,5′-[3-(2-Pyridyl)-1,2,4-triazine-5,6-diyl]difuran-2-sulfonic acid disodium salt
Empirical Formula (Hill Notation):
C16H8N4Na2O8S2
CAS Number:
Molecular Weight:
494.37
Products
Product Category
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Special Grade
Technique