
(2S)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide

Empirical Formula (Hill Notation):
C14H20N2O
CAS Number:
Molecular Weight:
232.32
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1