
2-(Methacryloyloxy)ethyl acetoacetate

Synonym(s):
(2-Acetoacetoxy)ethyl methacrylate, Ethylene glycol monoacetoacetate monomethacrylate
Linear Formula:
CH3COCH2CO2CH2CH2O2CC(CH3)=CH2
CAS Number:
Molecular Weight:
214.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2