Skip to Content
Merck
2-Butanone suitable for HPLC, ≥99.7%

2-Butanone

2-Butanone suitable for HPLC, ≥99.7%
Synonym(s):
Ethyl methyl ketone, MEK, Methyl ethyl ketone, 2-Butanone
Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing