Skip to Content
Merck
1H,1H,2H-Perfluoro-1-hexene 99%

1H,1H,2H-Perfluoro-1-hexene

1H,1H,2H-Perfluoro-1-hexene 99%
Synonym(s):
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexene
Linear Formula:
CH2=CH(CF2)3CF3
CAS Number:
Molecular Weight:
246.07
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing