Skip to Content
BT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1-Chlorobutane
Synonym(s):
Butyl chloride, 1-Chlorobutane
Linear Formula:
CH
3
(CH
2
)
3
Cl
CAS Number:
109-69-3
Molecular Weight:
92.57
EC Number:
203-696-6
Products
Product Category
solvents (4)
building blocks (2)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Anhydrous (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
34958
1-Chlorobutane,
suitable for HPLC, ≥99.8%
Expand
View Pricing
1.01692
1-Chlorobutane,
for liquid chromatography LiChrosolv
®
Expand
View Pricing
125008
1-Chlorobutane,
ReagentPlus
®
, 99%
Expand
View Pricing
8.01640
1-Chlorobutane,
for synthesis
Expand
View Pricing
414255
1-Chlorobutane,
anhydrous, 99.5%
Expand
View Pricing
09651
1-Chlorobutane,
biotech. grade, for protein sequence analysis, ≥99.8%
Expand
View Pricing