
1α,25-Dihydroxyvitamin D3

Synonym(s):
1α,25-Dihydroxycholecalciferol, Calcitriol
Empirical Formula (Hill Notation):
C27H44O3
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing