
1-(2-Methoxyphenyl)piperazine

Empirical Formula (Hill Notation):
C11H16N2O
Products
Product Category
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing