
1,1-Bis(methylthio)-2-nitroethylene

Synonym(s):
Nitroketene dimethyl mercaptal
Linear Formula:
O2NCH=C(SCH3)2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing