Milli-Q® Lab Water Purification Consumables
Our specialized portfolio of lab water system consumables covers every stage of the water purification process—from pretreatment and purification, to storage and distribution. Use the table below to find the cartridges, UV lamps, filters, and polishers you need for your water purification system.
Advantages of Milli-Q® water purification consumables include:
- Adaptable to different feed water qualities
- Consistent and reliable water quality for almost any application
- Validated with strict quality assurance protocols
- Genuine ISO 9001 certified parts
- Designed to perfectly fit your Milli-Q® water purification system
- Reduced end-of-life impact of purification cartridges with the Drain Cap
Popular Categories
Guarantee Water Quality & Protect Your System with Genuine Parts
Purification cartridges and other consumables play a core role in the water purification process as their objective is to remove or keep out contaminants. This protects the quality of your purified water supply and prevents system damage.
To ensure consistently high-quality purified water and the long lifetime of your water purification system, it’s critical to replace your water system consumables on time and with genuine ISO 9001-certified parts. On-time replacement of water system consumables:
- Helps to guarantee water quality
- Prevents system damage and contamination
- Fulfills validation criteria
- Protects the integrity of your lab’s work
Adapt water quality with purification cartiridges & final filters
NEW! Cartridges are now tailored to your pure water feed
IPAK Meta® and IPAK Quanta® polishing cartridges are now tailored to the quality of your pure feed water. For Milli-Q® IQ and EQ systems fed by…
- Elix® pure water, the resin mix has been reduced with no impact on performance. Cartridges are still exchanged once a year.
- Non-Elix® pure water, the resin mix has been modified to ensure the consistent quality of ultrapure water under more difficult feed water conditions.
Application POD-Paks match water quality to your science
Does your lab use purified water to perform a variety of applications? You'll find that our range of application POD-Paks is the perfect solution to meet the needs of your various applications and your most stringent requirements. Each POD-Pak filter or polisher targets specific types of contaminants and removes them right at the Q-POD® or E-POD® dispenser. If you need several types of ultrapure or pure water, simply connect different POD-Paks to your different dispensing units.
The POD-Pak range includes:
- Millipak® & sterile Millipak® Gold 0.22 µm filters: Bacteria- and particulate-free water
- Biopak® polisher: Pyrogen-, nuclease-, protease- and bacteria-free water
- VOC-Pak® polisher: water for volatile organic compounds related analyses
- EDS-Pak® polisher: water for endocrine disruptor-related applications
- LC-Pak® polisher: water for ultra-trace organic sensitive applications
Our cartridges are designed with sustainability in mind, reducing their end-of-life impact and helping labs meet their sustainability targets.
- Reduce Disposal Weight: The innovative Drain Cap purges trapped water from exhausted purification cartridges (IPAK Gard®, IPAK Meta®, IPAK Quanta®), reducing disposal weight by up to 35%. Watch the video to see how it works.
- Eliminate Mercury Handling: Our zero-mercury ech2o® UV lamps, used for bactericidal control and photo-oxidation of organic contaminants, reduce regulated waste management. They are also more compact than previous UV lamps, allowing for a smaller system.
- Greener Packaging: Cartridges are packed in 100% recycled cardboard with sustainable forestry certification. Redesigned cartridge kits reduce packaging weight by 51% and volume by 45%.
Never risk water quality with a Consumables Supply Agreement
Keep your Milli-Q® system running at peak performance and make ordering easy. A Consumables Supply Agreement (CSA) ensures you always have the right consumables at the right time—plus it simplifies the purchasing process. It’s the easiest, most economical and reliable solution to guarantee your water’s quality.
- Only 1 purchase order
- Automatic deliveries
- Never miss an expiry date
- Preferred pricing
- Flexibility to adjust
- Only genuine ISO 9001-certified parts from traceable lots
Ensure data traceability with e-Sure RFID tags
e-Sure tags provide indispensable data traceability. Based on RFID technology, these digital markings are present on most consumables to provide quality and safety assurance, as well as traceability.
- Automatically record consumables data and events in system memory
- Monitor consumable status directly on the system interface
Related Product Resources
- Flyer: Consumables Supply Agreement
- Application Note: Endotoxin-Free Water for Tissue Culture
- Datasheet: Application Pak Point-of-Use Polishers
- Datasheet: EDS-Pak® Polisher
- Water for Nuclease-sensitive Molecular Biology Studies
Nuclease-free water (or PCR water) is needed in molecular biology to avoid sample degradation during PCR, sequencing, etc. Ultrafiltration provides a quicker, safer, and more sustainable alternative to DEPC treatment when preparing RNase-free and DNase-free water.
- Nuclease-Free Water at Your Fingertips
An ultrafiltration cartridge can be placed at the outlet of a water purification system to deliver nuclease-free ultrapure water.