Skip to Content
Merck
All Photos(1)

Key Documents

SAB1409914

Sigma-Aldrich

Anti-RHPN2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

RhoBP, p76RBE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 77 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHPN2(85415)

General description

This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton. (provided by RefSeq)

Immunogen

RHPN2 (NP_149094.2, 1 a.a. ~ 686 a.a) full-length human protein.

Sequence
MTDALLPAAPQPLEKKNDGYFRKGCNPLAQTGRSKLQNQRAALNQQILKAVRMRTGAENLLKVATNSKVREQVRLELSFVNSDLQMLKEELEGLNISVGVYQNTEEAFTIPLIPLGLKETKDVDFAVVLKDFILEHYSEDGYLYEDEIADLMDLRQACRTPSRDEAGVELLMTYFIQLGFVESRFFPPTRQMGLLFTWYDSLTGVPVSQQNLLLEKASVLFNTGALYTQIGTRCDRQTQAGLESAIDAFQRAAGVLNYLKDTFTHTPSYDMSPAMLSVLVKMMLAQAQESVFEKISLPGIRNEFFMLVKVAQEAAKVGEVYQQLHAAMSQAPVKENIPYSWASLACVKAHHYAALAHYFTAILLIDHQVKPGTDLDHQEKCLSQLYDHMPEGLTPLATLKNDQQRRQLGKSHLRRAMAHHEESVREASLCKKLRSIEVLQKVLCAAQERSRLTYAQHQEEDDLLNLIDAPSVVAKTEQEVDIILPQFSKLTVTDFFQKLGPLSVFSANKRWTPPRSIRFTAEEGDLGFTLRGNAPVQVHFLDPYCSASVAGAREGDYIVSIQLVDCKWLTLSEVMKLLKSFGEDEIEMKVVSLLDSTSSMHNKSATYSVGMQKTYSMICLAIDDDDKTDKTKKISKKLSFLSWGTNKNRQKSASTLCLPSVGAARPQVKKKLPSPFSLLNSDSSWY

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service