GE17-0852-01
NAP™-25 20 ST
Cytiva 17-0852-01, pkg of 20 columns
Synonym(s):
20 ST NAP-25, NAP-25 Column, NAP-25 Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105501
NACRES:
NA.52
Recommended Products
packaging
pkg of 20 columns
manufacturer/tradename
Cytiva 17-0852-01
storage condition
(.01 M NaOH)
particle size
20-80 μm (dry)
working range
2-13
capacity
<1 mL binding capacity
General description
Disposable columns prepacked with Sephadex® G-25 DNA Grade and require only gravity to run.
Application
NAP™ Columns are disposable columns prepacked with Sephadex® G-25 DNA Grade and require only gravity to run. Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration, washing and sample elution.
Features and Benefits
- For rapid and efficient purification of DNA and oligonucleotides (10-mers) in less than 15 min by gravity flow.
- Prepacked with Sephadex® G-25 DNA Grade in distilled water containing 0.15% Kathon CG/ICP Biocide.
- Available in three sizes depending on sample volume: 0.5 mL (NAP-5), 1 mL (NAP-10) or 2.5 mL (NAP-25).
- Useful for small-scale purification, desalting and buffer exchange.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
NAP is a trademark of Cytiva
Sephadex is a registered trademark of Cytiva
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service