Skip to Content
Merck
All Photos(1)

Key Documents

EP0030501101

Eppendorf® Deepwell plates, standard, 96 wells

conical bottom colorless wells, working volume 500 μL, white plate

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15

material

conical bottom colorless wells
white plate

sterility

non-sterile

manufacturer/tradename

Eppendorf® 0030501101

availability

available only in (India)

technique(s)

PCR: suitable

size

96 wells

working volume

500 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® deepwell plates are manufactured from chemically resistant polypropylene (PP)
  • Ideal for most manual and automated applications.
  • Features a high-contrast laser-engraved alphanumeric label to identify wells.
  • OptiTrack® matrix facilitates up to 30 % faster sample identification.
  • RecoverMax® well design for optimized well geometry that enables maximum sample recovery.
  • Deepwell plates are free of detectable slip agents, plasticizers, and biocides for contamination-free bioassays.
  • Autoclavable (121°C, 20 min).
For greater reliability, stability and efficiency, choose Eppendorf Deepwell 96 well Plates. Available with white, yellow, green or blue frames, these deepwell plates offer high performance for all applications - both manual and automated. Thanks to OptiTrack matrix, a high-contrast alphanumeric label laser-engraved into the frame, you can find your sample up to 30% faster and reduce pipetting errors. For every application, from sample storage at 86 C to DNA denaturation at 100 C, rely on Eppendorf Deepwell Plates and set new standards in your laboratory.

Application

  • Sample storage and preparation Bacteria and yeast cultivation
  • High-throughput DNA and RNA isolation
  • Storage of cDNA or genomic libraries
  • Storage of siRNA or oligonucleotide libraries
  • Protein analysis Storage of valuable protein and DNA samples in Eppendorf LoBind Plates
  • High-throughput plasmid purification Creation of dilution series

Features and Benefits

  • OptiTrack matrix: up to 30 % faster sample detection and fewer pipetting errors thanks to high-contrast alphanumeric labeling
  • RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • g-Safe : exceptional centrifugation resistance up to 6,000 × g for faster protocols and improved sample quality
  • Raised corrugated edges and smooth surface for reliable sealing, even during heat sealing
  • Easily and reliably stackable
  • Minimal residual volume and high homogeneity from well to well
  • High-quality polypropylene for high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without mold release agents, plasticizers and biocides – substances that can negatively affect bioassays
  • Autoclavable (121 C, 20 min)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service