Skip to Content
Merck
All Photos(1)

Documents

Vero C1008 [Vero 76, clone E6, Vero E6]

85020206, African green monkey kidney, Epithelial

Synonym(s):

Vero 76 Clone E6 Cells, Vero Cells, Vero E6 Cells, VeroC1008 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Vero C1008 [Vero 76, clone E6, Vero E6], 85020206, from African green monkey kidney, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

African green monkey kidney

growth mode

Adherent

karyotype

2n = 60

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Vero cells are derived from the kidney of an African green monkey. These are anchorage-dependent cells that have applications in molecular and cell biology research. Vero E6 cells enable achieving high titers of severe acute respiratory syndrome coronavirus (SARS-CoV virus) and are also used in the cultivation of Rabies virus, Reovirus and Japanese encephalitis virus.

Cell Line Origin

Monkey African Green kidney

Cell Line Description

A subclone of Vero76 (Sigma catalog no. 85020205) exhibiting a similar range of virus susceptibility (haemorrhagic viruses). They show some degree of contact inhibition and are suitable for supporting the growth of slowly replicating viruses.

Application

VERO C1008 has been used for transfection, cultivation and propagation of West Nile virus, yellow fever virus, ZIKA, Ebola virus, Sabethes flavivirus (SbFV), Crimean-Congo Haemorrhagic Fever (CCHF), Human-derived Lake Victoria Marburg virus strain Popp.

Culture Medium

DMEM (D6546) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service