Skip to Content
Merck
All Photos(1)

Key Documents

388M-1

Sigma-Aldrich

Nestin (10C2) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

10C2, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (388M-14)
vial of 0.5 mL concentrate (388M-15)
bottle of 1.0 mL predilute (388M-17)
vial of 1.0 mL concentrate (388M-16)
bottle of 7.0 mL predilute (388M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... NES(10763)

General description

Nestin is a class VI intermediate filament (IF) protein. It has been reported that nestin expression is significantly increased in melanoma and correlated with more advanced stages of the disease. An immunohistochemical analysis identified nestin-positive cells in 84% (35/42) of primary melanoma and 83% (10/12) of metastatic melanoma. Nestin immunoreactivity was also observed in melanoma cells in all (10/10) cases of HMB-45-negative amelanotic and melanotic, non-desmoplastic melanoma. Nestin expression has been reported in tumors of the CNS, including astrocytoma, ependymoma, oligodendroglioma, glioblastoma, and primitive neuroectodermal tumors. Nestin has been detected in human gliomas, including low and high grade, but its expression is observed more frequently in high grade than in low grade gliomas. Nestin overexpression is seen in carcinomas, including prostatic adenocarcinoma, pancreatic ductal carcinoma, thyroid carcinoma, and in mesenchymal tumors such as GIST and DFSP. Among the breast carcinoma subtypes, nestin is highly expressed in basal breast cancer subtype (ERα-/PR-/Her2-) but not in the Her2 subtype (ERα-/PR-/Her2+) or luminal epithelial phenotype (ERα+/PR+). Only cytoplasmic staining is considered positive, whereas any nuclear staining is considered as background artifact. In normal skin, nestin is observed in endothelial cells and the bulge area of hair follicles.

Quality


IVD

IVD

IVD

RUO

Linkage

Nestin Positive Control Slides, Product No. 388S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiyuki Ishiwata et al.
World journal of gastroenterology, 17(4), 409-418 (2011-01-29)
Nestin is a class VI intermediate filament protein that was originally described as a neuronal stem cell marker during central nervous system (CNS) development, and is currently widely used in that capacity. Nestin is also expressed in non-neuronal immature or
Svetlana Brychtova et al.
Journal of cutaneous pathology, 34(5), 370-375 (2007-04-24)
Nestin is one of the intermediate filaments that are expressed in proliferating neural progenitor cells during development of the central nervous system (CNS) and peripheral nervous system. Postnatal re-expression of the protein occurs mainly under pathological conditions, including injury and
Hua Li et al.
Cancer research, 67(2), 501-510 (2007-01-20)
Transcriptional profiling has identified five breast cancer subtypes, of which the basal epithelial is most aggressive and correlates with poor prognosis. These tumors display a high degree of cellular heterogeneity and lack established molecular targets, such as estrogen receptor-alpha, progesterone
Maho Kanoh et al.
The Journal of dermatology, 37(6), 505-511 (2010-06-12)
Nestin, a marker of neural stem cells, is expressed in the stem cells of the mouse hair follicle. The nestin-expressing hair follicle stem cells can differentiate into neurons, glia, keratocytes, smooth muscle cells and melanocytes in vitro. These pluripotent nestin-expressing

Articles

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service