Skip to Content
Merck
All Photos(1)

Key Documents

67996

Sigma-Aldrich

Tris Acetate-EDTA buffer Concentrate

for Northern and Southern blotting, solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

grade

for molecular biology

form

solution

quality

50x Concentrate

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

8.3±0.2 (25 °C)

density

1.09 g/mL at 20 °C

storage temp.

room temp

General description

TBE (Tris/Acetate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. This solution has a lower buffering capacity than TBE buffer, but double stranded DNA runs faster with TAE buffer. This 50x concentrate can be easily diluted with molecular biology grade water to 1x before use.

Application

Tris Acetate-EDTA buffer concentrate is suitable for use as running buffer for nucleic acid electrophoresis, which may be part of Northern and Southern blot hybridization procedures. It is especially useful for double stranded DNA separations.

Components

The TAE 50x liquid concentrate contains 2M TRIS acetate and 0.05M EDTA in water.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service